ENA300791454 Enamine

5-[4-(propan-2-yl)phenyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD11201451
  • Cas Number : 1038309-85-1
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66871
Availability details for
Quantity
  • Availability
  • Add to Cart