ENA300791457 Enamine

N'-(2-chloroacetyl)-3-methylbenzohydrazide

  • MDL number MFCD04545070
  • Cas Number : 862853-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18179

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18179
Availability details for
Quantity
  • Availability
  • Add to Cart