ENA300791459 Enamine

[3-(dimethylamino)-2,2-dimethylpropyl]thiourea

  • MDL number MFCD11202356
  • Cas Number : 1038374-89-8
  • Molecular Weight : 189.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66876
Availability details for
Quantity
  • Availability
  • Add to Cart