ENA300791461 Enamine

3-(aminomethyl)-N-[2-(dimethylamino)ethyl]benzamide

  • MDL number MFCD09044372
  • Cas Number : 926198-12-1
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66880
Availability details for
Quantity
  • Availability
  • Add to Cart