ENA300791463 Enamine

N-ethylpyridin-3-amine

  • MDL number MFCD11140216
  • Cas Number : 32405-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66883

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P311,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-66883
pictograms GHS06
hstatements H302,H315,H319,H331,H335
Availability details for
Quantity
  • Availability
  • Add to Cart