ENA300791468 Enamine

2-methyl-N-(piperidin-4-yl)furan-3-carboxamide

  • MDL number MFCD09731347
  • Cas Number : 953733-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66890
Availability details for
Quantity
  • Availability
  • Add to Cart