ENA300791474 Enamine

1-(1-chloroethyl)-3-fluorobenzene

  • MDL number MFCD10691958
  • Cas Number : 58114-09-3
  • Molecular Weight : 158.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66896
Availability details for
Quantity
  • Availability
  • Add to Cart