ENA300791476 Enamine

2-fluoro-5-(5-methyl-1H-1,2,3,4-tetrazol-1-yl)aniline

  • MDL number MFCD12091833
  • Cas Number : 1156374-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-66898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66898
Availability details for
Quantity
  • Availability
  • Add to Cart