ENA300791479 Enamine

1-ethyl-2-methyl-4,5,6,7-tetrahydro-1H-indol-4-amine

  • MDL number MFCD16150168
  • Cas Number : 1249427-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-63463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-63463
Availability details for
Quantity
  • Availability
  • Add to Cart