ENA300791490 Enamine

6-(4-methoxyphenyl)hexanoic acid

  • MDL number MFCD12172613
  • Cas Number : 107228-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46...48
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66919

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-66919
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart