ENA300791492 Enamine

1-[4-methyl-2-(4-methylphenyl)-1,3-thiazol-5-yl]ethan-1-one

  • MDL number MFCD07354132
  • Cas Number : 54001-03-5
  • Molecular Weight : 231.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66921
Availability details for
Quantity
  • Availability
  • Add to Cart