ENA300791495 Enamine

2-fluoro-6-(pyrrolidin-1-yl)aniline

  • MDL number MFCD12149452
  • Cas Number : 1183840-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66925
Availability details for
Quantity
  • Availability
  • Add to Cart