ENA300791497 Enamine

2-methyl-2-[3-(trifluoromethyl)phenyl]oxirane

  • MDL number MFCD16817559
  • Cas Number : 850402-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66927
Availability details for
Quantity
  • Availability
  • Add to Cart