ENA300791500 Enamine

N-(propan-2-yl)-9H-purin-6-amine

  • MDL number MFCD04142660
  • Cas Number : 63752-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-63843
Availability details for
Quantity
  • Availability
  • Add to Cart