ENA300791502 Enamine

4-chloro-N-(4-methylphenyl)-3-nitrobenzamide

  • MDL number MFCD01006988
  • Cas Number : 328258-50-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66933
Availability details for
Quantity
  • Availability
  • Add to Cart