ENA300791504 Enamine

N-{2-[(4-hydroxyphenyl)sulfanyl]ethyl}acetamide

  • MDL number MFCD07779474
  • Cas Number : 91281-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66963
Availability details for
Quantity
  • Availability
  • Add to Cart