ENA300791506 Enamine

2-(3-bromopropoxy)naphthalene

  • MDL number MFCD02030535
  • Cas Number : 3245-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53-57
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-66965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66965
Availability details for
Quantity
  • Availability
  • Add to Cart