ENA300791508 Enamine

5-(4-propanoylphenoxy)pentanoic acid

  • MDL number MFCD11542180
  • Cas Number : 1039915-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66967

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66967
Availability details for
Quantity
  • Availability
  • Add to Cart