ENA300791510 Enamine

4-(dimethylamino)-3-fluorobenzaldehyde

  • MDL number MFCD11156408
  • Cas Number : 1021240-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66969

Safety Information

msds https://www.enaminestore.com/catalog/EN300-66969
Availability details for
Quantity
  • Availability
  • Add to Cart