ENA300791512 Enamine

N-(3-methylbutyl)-1,3-thiazolidine-4-carboxamide

  • MDL number MFCD09950484
  • Cas Number : 1104608-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66972
Availability details for
Quantity
  • Availability
  • Add to Cart