ENA300791516 Enamine

3,4-dichloro-N-(2-hydroxyphenyl)benzene-1-sulfonamide

  • MDL number MFCD07359413
  • Cas Number : 881302-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67011
Availability details for
Quantity
  • Availability
  • Add to Cart