ENA300791519 Enamine

3-phenyl-1,3-diazinane-2,4-dione

  • MDL number MFCD12137761
  • Cas Number : 5100-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67017
Availability details for
Quantity
  • Availability
  • Add to Cart