ENA300791520 Enamine

4-chloro-1,2,5-thiadiazol-3-ol

  • MDL number MFCD08235253
  • Cas Number : 5728-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-112
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67018
Availability details for
Quantity
  • Availability
  • Add to Cart