ENA300791521 Enamine

[3-(dimethylamino)propyl](diphenylmethyl)amine

  • MDL number MFCD12552669
  • Cas Number : 807277-01-6
  • Molecular Weight : 268.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67020
Availability details for
Quantity
  • Availability
  • Add to Cart