ENA300791525 Enamine

N-(2,4-dimethylphenyl)-1,3-thiazolidine-4-carboxamide

  • MDL number MFCD09929399
  • Cas Number : 1104793-76-1
  • Molecular Weight : 236.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2,4-dimethylphenyl)-1,3-thiazolidine-4-carboxamide
Inchi Code 1S/C12H16N2OS/c1-8-3-4-10(9(2)5-8)14-12(15)11-6-16-7-13-11/h3-5,11,13H,6-7H2,1-2H3,(H,14,15)
Inchi Key WJYKZASSPVLZDZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67026
Availability details for
Quantity
  • Availability
  • Add to Cart