ENA300791531 Enamine

1-{4-[(3-methylphenyl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD02270860
  • Cas Number : 890092-06-5
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67032
Availability details for
Quantity
  • Availability
  • Add to Cart