ENA300791532 Enamine

1-{[3-(trifluoromethyl)phenyl]methyl}-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD13465389
  • Cas Number : 1038711-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67033
Availability details for
Quantity
  • Availability
  • Add to Cart