ENA300791536 Enamine

1,1-dioxo-1lambda6-thiomorpholine-4-carbothioamide

  • MDL number MFCD12142626
  • Cas Number : 21428-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67039
Availability details for
Quantity
  • Availability
  • Add to Cart