ENA300791538 Enamine

N-(4-fluorophenyl)piperidine-4-carboxamide hydrochloride

  • MDL number MFCD13562831
  • Cas Number : 1220038-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-64265
Availability details for
Quantity
  • Availability
  • Add to Cart