ENA300791539 Enamine

1$l^{6},4-thiomorpholine-1,1-dione hydrochloride

  • MDL number MFCD06738995
  • Cas Number : 59801-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 247...249
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67050

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-67050
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart