ENA300791546 Enamine

4-(butan-2-yl)-1,3-thiazol-2-amine

  • MDL number MFCD12767718
  • Cas Number : 79932-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67058
Availability details for
Quantity
  • Availability
  • Add to Cart