ENA300791550 Enamine

3-(2,3-dimethylphenyl)-3-oxopropanenitrile

  • MDL number MFCD02260822
  • Cas Number : 898787-06-9
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67065
Availability details for
Quantity
  • Availability
  • Add to Cart