ENA300791551 Enamine

3-(2,4-dinitrophenyl)propanamide

  • MDL number MFCD01110993
  • Cas Number : 681466-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67066

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67066
Availability details for
Quantity
  • Availability
  • Add to Cart