ENA300791553 Enamine

4-tert-butyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00120352
  • Cas Number : 29983-27-5
  • Molecular Weight : 157.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67068
Availability details for
Quantity
  • Availability
  • Add to Cart