ENA300791556 Enamine

methyl 2-(3-amino-1H-pyrazol-1-yl)acetate

  • MDL number MFCD12149369
  • Cas Number : 1093967-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74...76
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67072

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67072
Availability details for
Quantity
  • Availability
  • Add to Cart