ENA300791557 Enamine

2-(2-chloro-6-fluorophenyl)-3-oxobutanenitrile

  • MDL number MFCD00055280
  • Cas Number : 93839-17-9
  • Molecular Weight : 211.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67074
Availability details for
Quantity
  • Availability
  • Add to Cart