ENA300791567 Enamine

1-(2,6-difluorobenzoyl)piperazine

  • MDL number MFCD09802353
  • Cas Number : 179334-15-7
  • Molecular Weight : 226.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67090
Availability details for
Quantity
  • Availability
  • Add to Cart