ENA300791571 Enamine

3-pentanoyl-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD09930464
  • Cas Number : 1105692-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67095
Availability details for
Quantity
  • Availability
  • Add to Cart