ENA300791587 Enamine

N,N-dimethyl-2,3-dihydro-1H-indole-5-carboxamide

  • MDL number MFCD11181066
  • Cas Number : 1094284-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67129
Availability details for
Quantity
  • Availability
  • Add to Cart