ENA300791589 Enamine

1-(2,2,2-trifluoroethyl)-1H-imidazole-2-carbaldehyde

  • MDL number MFCD09908506
  • Cas Number : 497855-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67131
Availability details for
Quantity
  • Availability
  • Add to Cart