ENA300791592 Enamine

N-(1-{4-[(4-fluorophenyl)methoxy]phenyl}ethylidene)hydroxylamine

  • Cas Number : 1020973-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67136
Availability details for
Quantity
  • Availability
  • Add to Cart