ENA300791596 Enamine

6-[(3-aminophenoxy)methyl]-N2,N2-dimethyl-1,3,5-triazine-2,4-diamine

  • MDL number MFCD12856647
  • Cas Number : 1183585-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67142
Availability details for
Quantity
  • Availability
  • Add to Cart