ENA300791597 Enamine

1-bromo-4-[(propan-2-ylsulfanyl)methyl]benzene

  • MDL number MFCD14644602
  • Cas Number : 1249091-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67143
Availability details for
Quantity
  • Availability
  • Add to Cart