ENA300791603 Enamine

3-methylnonanoic acid

  • MDL number MFCD02258980
  • Cas Number : 35205-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-67149

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67149
Availability details for
Quantity
  • Availability
  • Add to Cart