ENA300791610 Enamine

4-methyl-2-(2-methylpropyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD11207789
  • Cas Number : 1094231-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67159
Availability details for
Quantity
  • Availability
  • Add to Cart