ENA300791615 Enamine

3-methanesulfonyl-4H-1,2,4-triazole

  • MDL number MFCD01422981

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-65164

Safety Information

msds https://www.enaminestore.com/catalog/EN300-65164
Availability details for
Quantity
  • Availability
  • Add to Cart