ENA300791616 Enamine

ethyl 6-sulfanylpyridine-3-carboxylate

  • MDL number MFCD09733239
  • Cas Number : 175909-91-8
  • Molecular Weight : 183.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67169
Availability details for
Quantity
  • Availability
  • Add to Cart