ENA300791624 Enamine

N-(cyclopropylmethyl)-2-(methylamino)ethane-1-sulfonamide

  • MDL number MFCD14688117
  • Cas Number : 1250417-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67178

Safety Information

msds https://www.enaminestore.com/catalog/EN300-67178
Availability details for
Quantity
  • Availability
  • Add to Cart