ENA300791626 Enamine

2-(3-methylbutyl)cyclopentan-1-amine

  • MDL number MFCD14653494
  • Cas Number : 1248231-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67180

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67180
Availability details for
Quantity
  • Availability
  • Add to Cart