ENA300791627 Enamine

9-chloro-2-methyl-2-azatricyclo[6.3.1.0,4,12]dodeca-1(11),4(12),5,7,9-pentaen-3-one

  • MDL number MFCD00604757
  • Cas Number : 39158-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18655
Availability details for
Quantity
  • Availability
  • Add to Cart